Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Immp2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053122.4 | NP_444352.2 | 754 | |||
GenBank mRNA | AF359564.1 | - | 686 | |||
AK053361.1 | - | 728 | ||||
AK161351.1 | - | 894 | ||||
BB819807.1 | - | 1 | 17 | |||
BC061049.1 | - | 754 | ||||
BC100557.1 | - | 706 | 5057188 |