Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarca5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053124.2 | NP_444354.2 | 23 | 3367 | ||
GenBank mRNA | AF325921.1 | - | 23 | 3019 | ||
AF375046.1 | - | 23 | 3086 | |||
AK052320.1 | - | 11 | 1320 | |||
AK156915.1 | - | 7 | 831 | |||
BC021922.1 | - | 11 | 1305 | |||
BC053069.1 | - | 23 | 3367 | 6409140 |