Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhb13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053138.2 | NP_444368.1 | 1 | 1196 | ||
GenBank mRNA | AK082435.1 | - | 1 | 1203 | ||
AY013774.1 | - | 1 | 1142 | |||
BC138123.1 | - | 1 | 1213 | 8861142 | ||
BC138124.1 | - | 1 | 1213 | 8861143 |