Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Clmn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040682.1 | NP_001035772.1 | 10 | 2806 | ||
NM_053155.2 | NP_444385.2 | 10 | 2806 | |||
GenBank mRNA | AB047978.1 | - | 10 | 2799 | ||
AB059643.1 | - | 10 | 2799 | |||
AB059644.1 | - | 10 | 2799 | |||
AB059645.1 | - | 10 | 2799 | |||
AB059646.1 | - | 10 | 2799 | |||
AB059647.1 | - | 10 | 2799 | |||
AB059648.1 | - | 10 | 2799 | |||
AK077023.1 | - | 7 | 2388 | |||
AK220261.1 | - | 2 | 785 | |||
BC141280.1 | - | 10 | 2739 | 9055892 |