Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcoln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053177.1 | NP_444407.1 | 3, 4 | 488 | ||
GenBank mRNA | AF302009.1 | - | 3, 4 | 485 | ||
AK028385.1 | - | 3, 4 | 465 | |||
AK034471.1 | - | 3, 4 | 499 | |||
AK038594.1 | - | 3, 4 | 501 | |||
AK049606.1 | - | 3, 4 | 489 | |||
BC005651.1 | - | 3, 4 | 488 | 3257440 | ||
BC020377.1 | - | 2, 3 | 338 |