Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcoln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053177.1 | NP_444407.1 | 9, 10 | 1223 | ||
GenBank mRNA | AF302009.1 | - | 9, 10 | 1220 | ||
AK028385.1 | - | 9, 10 | 1200 | |||
AK034471.1 | - | 9, 10 | 1234 | |||
AK038594.1 | - | 9, 10 | 1236 | |||
AK049606.1 | - | 9, 10 | 1224 | |||
BC005651.1 | - | 9, 10 | 1223 | 3257440 | ||
BC020377.1 | - | 6 | 1665 |