Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRMT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039619.2 | NP_001034708.1 | 7 | 1040 | ||
NM_001282953.1 | NP_001269882.1 | 6 | 908 | |||
NM_001282954.1 | NP_001269883.1 | 6 | 921 | |||
NM_001282955.1 | NP_001269884.1 | 6 | 823 | |||
NM_006109.4 | NP_006100.2 | 7 | 955 | |||
XM_017020927.1 | XP_016876416.1 | 6 | 679 | |||
GenBank mRNA | AB451246.1 | - | 7 | 731 | ||
AB451370.1 | - | 7 | 731 | |||
AF167572.1 | - | 7 | 822 | |||
AK075251.1 | - | 7 | 833 | |||
AK300863.1 | - | 6 | 705 | |||
AK301812.1 | - | 6 | 714 | |||
AK302240.1 | - | 6 | 603 | |||
AK315500.1 | - | 7 | 771 | |||
AL833612.1 | - | 7 | 740 | |||
BC025979.2 | - | 7 | 743 | 3833019 | ||
CR456741.1 | - | 7 | 731 |