Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfand2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159908.1 | NP_001153380.1 | 7 | 1066 | ||
NM_133349.3 | NP_579927.1 | 7 | 1077 | |||
GenBank mRNA | AF224494.1 | - | 7 | 963 | ||
AK146082.1 | - | 7 | 1015 | |||
AK171245.1 | - | 7 | 1017 | |||
AK189370.1 | - | 1 | 234 | |||
BC058780.1 | - | 7 | 1003 | 6312202 | ||
BU840945.1 | - | 3 | 439 |