Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfand2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159908.1 | NP_001153380.1 | 7 | 2411 | ||
NM_133349.3 | NP_579927.1 | 7 | 2422 | |||
GenBank mRNA | AF224494.1 | - | 7 | 2308 | ||
AK146082.1 | - | 7 | 2362 | |||
AK219098.1 | - | 2 | 262 | |||
BC058780.1 | - | 7 | 2346 | 6312202 |