Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRAME | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291715.1 | NP_001278644.1 | 5 | 1981 | ||
NM_001291716.1 | NP_001278645.1 | 5 | 1960 | |||
NM_001291717.1 | NP_001278646.1 | 5 | 2079 | |||
NM_001291719.1 | NP_001278648.1 | 4 | 1862 | |||
NM_001318126.1 | NP_001305055.1 | 4 | 1743 | |||
NM_001318127.1 | NP_001305056.1 | 4 | 2005 | |||
NM_006115.4 | NP_006106.1 | 6 | 1994 | |||
NM_206953.2 | NP_996836.1 | 5 | 2608 | |||
NM_206954.2 | NP_996837.1 | 6 | 1973 | |||
NM_206955.2 | NP_996838.1 | 6 | 2198 | |||
NM_206956.2 | NP_996839.1 | 6 | 2177 | |||
XM_011530034.2 | XP_011528336.1 | 5 | 2279 | |||
GenBank mRNA | AF025440.1 | - | 1 | 320 | ||
AK129783.1 | - | 5 | 1845 | |||
AK302188.1 | - | 4 | 2004 | |||
AK304593.1 | - | 5 | 2004 | |||
AK316395.1 | - | 4 | 1743 | |||
BC014074.2 | - | 6 | 1831 | 3954524 | ||
BC022008.1 | - | 5 | 2619 | 4825127 | ||
BC039731.1 | - | 5 | 2502 | 5744328 | ||
U65011.1 | - | 6 | 1908 |