Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039878.2 | NP_001034967.1 | 17 | 2592 | ||
NM_133789.3 | NP_598550.2 | 17 | 2613 | |||
GenBank mRNA | AF414080.1 | - | 17 | 2613 | ||
AK042242.1 | - | 3 | 225 | |||
AK136155.1 | - | 17 | 1909 | |||
AK154615.1 | - | 17 | 2142 | |||
AK161673.1 | - | 17 | 2164 | |||
BC004025.1 | - | 15 | 1726 | |||
BC022667.1 | - | 5 | 425 | |||
BC066805.1 | - | 4 | 2955 | |||
BC080283.1 | - | 17 | 2179 | 6401782 |