Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4833439L19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252645.1 | NP_001239574.1 | 5, 6 | 739 | ||
NM_001252646.1 | NP_001239575.1 | 4, 5 | 632 | |||
NM_001252647.1 | NP_001239576.1 | 3, 4 | 453 | |||
NM_001252648.1 | NP_001239577.1 | 2, 3 | 326 | |||
NM_133797.4 | NP_598558.1 | 6, 7 | 827 | |||
GenBank mRNA | AK004845.1 | - | 6, 7 | 786 | ||
AK014792.1 | - | 2, 3 | 260 | |||
AK028697.1 | - | 6, 7 | 760 | |||
AK145025.1 | - | 6, 7 | 793 | |||
AK159945.1 | - | 6, 7 | 790 | |||
AK167616.1 | - | 6, 7 | 771 | |||
AK169399.1 | - | 6, 7 | 793 | |||
AK173126.1 | - | 6, 7 | 740 | |||
BC033445.1 | - | 6, 7 | 806 | 4222839 | ||
BC039775.1 | - | 6, 7 | 765 | 4239147 | ||
BC043040.1 | - | 6, 7 | 783 | 5701249 | ||
BC054417.1 | - | 6, 7 | 772 | 3969623 | ||
BY244215.1 | - | 3, 4 | 416 | |||
BY265900.1 | - | 2, 3 | 327 | |||
CA461006.1 | - | 4, 5 | 634 |