Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eps8l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133867.2 | NP_598628.1 | 6, 7 | 619 | ||
GenBank mRNA | AK136584.1 | - | 6, 7 | 615 | ||
AK167630.1 | - | 6, 7 | 606 | |||
AY074932.1 | - | 6, 7 | 635 | |||
BC014734.1 | - | 6, 7 | 607 | 4218079 |