Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134009.3 | NP_598770.1 | 11 | 1452 | ||
GenBank mRNA | AK019409.1 | - | 11 | 1322 | ||
AK048954.1 | - | 11 | 1446 | |||
AK128887.1 | - | 11 | 1453 | |||
AK206232.1 | - | 3 | 97 | |||
BC019501.1 | - | 11 | 1452 | 4193809 |