Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134033.2 | NP_598794.2 | 3 | 517 | ||
GenBank mRNA | BC018601.1 | - | 3 | 487 | ||
BC034558.1 | - | 3 | 455 | 3676182 | ||
BC059888.1 | - | 3 | 472 | 6816177 | ||
BQ768997.1 | - | 3 | 517 |