Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134033.2 | NP_598794.2 | 5 | 1664 | ||
GenBank mRNA | AK003430.1 | - | 1 | 247 | ||
BC003203.1 | - | 3 | 1092 | |||
BC008541.1 | - | 2 | 930 | |||
BC018601.1 | - | 5 | 1634 | |||
BC034558.1 | - | 5 | 1607 | 3676182 | ||
BC059888.1 | - | 5 | 1619 | 6816177 |