Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnxl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144868.3 | NP_659117.2 | 20 | 4115 | ||
GenBank mRNA | AF237953.1 | - | 18 | 2132 | ||
BC025838.1 | - | 14 | 1618 | |||
BC038056.1 | - | 15 | 2223 | |||
BC054751.1 | - | 20 | 3394 | |||
BC055289.1 | - | 21 | 3918 | |||
BC096538.1 | - | 20 | 4115 |