Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153419.2 | NP_700468.2 | 7 | 1454 | ||
GenBank mRNA | AB001539.1 | - | 8 | 1526 | ||
AK077611.1 | - | 7 | 1459 | |||
BC008121.1 | - | 7 | 1431 | |||
BC028896.1 | - | 6 | 1323 | 3674738 | ||
BC083143.1 | - | 7 | 1442 | |||
DT904438.1 | - | 1 | 269 |