Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Celf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289613.1 | NP_001276542.1 | 13 | 2580 | ||
NM_001289616.1 | NP_001276545.1 | 14 | 1978 | |||
NM_001289620.1 | NP_001276549.1 | 13 | 1921 | |||
NM_172434.3 | NP_766022.1 | 13 | 2493 | |||
GenBank mRNA | AK019578.1 | - | 10 | 2404 | ||
AK030259.1 | - | 12 | 1909 | |||
AK044506.1 | - | 14 | 1942 | |||
AK087536.1 | - | 13 | 1883 | |||
AY165052.1 | - | 13 | 2211 | |||
BC031740.1 | - | 9 | 2841 | |||
BC057553.1 | - | 13 | 2635 | 6401356 |