Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc41a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173865.3 | NP_776290.1 | 2 | 1274 | ||
GenBank mRNA | AK089802.1 | - | 2 | 1275 | ||
AK154819.1 | - | 2 | 1247 | |||
AK192565.1 | - | 1 | 16 | |||
BC096596.1 | - | 2 | 1310 | 4239523 | ||
BC116280.1 | - | 2 | 1082 | 40094408 | ||
BC116281.1 | - | 2 | 1082 | 40094411 |