Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034856.1 | - | 11, 12 | 1562 | ||
AK052103.1 | - | 9, 10 | 1290 | |||
AK085336.1 | - | 10, 11 | 1436 | |||
AK086462.1 | - | 11, 12 | 1594 | |||
BC054120.1 | - | 11, 12 | 1547 | 6506658 |