Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178877.6 | NP_849208.4 | 10, 11 | 1383 | ||
GenBank mRNA | AK029417.1 | - | 7, 8 | 934 | ||
AK039655.1 | - | 1, 2 | 382 | |||
BC090977.1 | - | 10, 11 | 1336 | 30503721 |