Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252458.1 | NP_001239387.1 | 15 | 2237 | ||
NM_178900.4 | NP_849231.1 | 14 | 2522 | |||
GenBank mRNA | AK037030.1 | - | 14 | 2523 | ||
AK154029.1 | - | 14 | 2371 | |||
BC095949.1 | - | 15 | 2187 | 4950957 | ||
BC096444.1 | - | 15 | 2205 | 4216827 |