Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atf7ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019426.2 | NP_062299.2 | 13 | 3413 | ||
GenBank mRNA | AJ132702.1 | - | 13 | 3406 | ||
AK132359.1 | - | 12 | 2033 | |||
AK137199.1 | - | 5 | 572 | |||
AK157479.1 | - | 3 | 1172 | |||
AK167542.1 | - | 14 | 3459 | |||
BC052460.1 | - | 13 | 3413 | 6834550 | ||
BC060246.1 | - | 13 | 3396 | 5705945 |