Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NCOR1 SIRNA Assays ›
Gene Symbol:
NCOR1
Gene Name:
nuclear receptor corepressor 1
Gene Aliases:
N-CoR, N-CoR1, PPP1R109, TRAC1, hN-CoR
Chromosome Location:
Chr.17: 16030094 - 16216746 on Build GRCh38
UniGene ID:
Hs.462323
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
17667
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NCOR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001190440.1
NP_001177369.1
2
278
NM_006311.3
NP_006302.2
3
477
XM_005256866.4
XP_005256923.1
3
460
XM_005256868.4
XP_005256925.1
3
460
XM_005256871.4
XP_005256928.1
3
460
XM_005256872.4
XP_005256929.1
3
460
XM_005256873.4
XP_005256930.1
3
460
XM_005256874.4
XP_005256931.1
3
460
XM_005256875.3
XP_005256932.1
3
460
XM_006721601.3
XP_006721664.1
4
592
XM_006721602.3
XP_006721665.1
4
590
XM_006721603.3
XP_006721666.1
3
460
XM_006721604.3
XP_006721667.1
3
460
XM_011524083.2
XP_011522385.1
3
1371
XM_011524084.2
XP_011522386.1
3
460
XM_011524085.2
XP_011522387.1
3
460
XM_011524086.2
XP_011522388.1
3
460
XM_017025396.1
XP_016880885.1
4
1337
XM_017025397.1
XP_016880886.1
4
1102
XM_017025398.1
XP_016880887.1
3
544
XM_017025399.1
XP_016880888.1
3
563
XM_017025400.1
XP_016880889.1
4
1337
XM_017025401.1
XP_016880890.1
4
1102
XM_017025402.1
XP_016880891.1
3
386
XM_017025403.1
XP_016880892.1
4
587
XM_017025404.1
XP_016880893.1
4
1078
XM_017025405.1
XP_016880894.1
3
973
XM_017025406.1
XP_016880895.1
3
1371
XM_017025407.1
XP_016880896.1
3
307
XM_017025408.1
XP_016880897.1
3
379
XM_017025409.1
XP_016880898.1
4
587
XM_017025410.1
XP_016880899.1
4
1083
XM_017025411.1
XP_016880900.1
3
826
XM_017025412.1
XP_016880901.1
3
603
XM_017025413.1
XP_016880902.1
3
386
XM_017025414.1
XP_016880903.1
2
293
XM_017025415.1
XP_016880904.1
3
460
XM_017025416.1
XP_016880905.1
4
1098
XM_017025417.1
XP_016880906.1
3
460
XM_017025418.1
XP_016880907.1
3
460
XM_017025419.1
XP_016880908.1
3
460
XM_017025420.1
XP_016880909.1
3
460
GenBank mRNA
AB028970.2
-
2
252
AF044209.1
-
3
448
AK307614.1
-
3
446
BC026028.1
-
4
551
BC050594.2
-
3
449
BC056862.1
-
3
393
BC058511.1
-
3
454
BC068996.1
-
3
449
BC107772.1
-
3
465
BC142648.1
-
3
431
BC167431.1
-
2
253
8860107
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001271103, BC100263.
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
skeletal muscle tissue development
lactation
circadian rhythm
positive regulation of cell proliferation
covalent chromatin modification
cerebellum development
cellular lipid metabolic process
negative regulation of JNK cascade
spindle assembly
regulation of glycolytic process by negative regulation of transcription from RNA polymerase II promoter
regulation of lipid transport by negative regulation of transcription from RNA polymerase II promoter
negative regulation of production of miRNAs involved in gene silencing by miRNA
regulation of fatty acid transport
RNA polymerase II regulatory region DNA binding
RNA polymerase II activating transcription factor binding
chromatin binding
transcription corepressor activity
protein binding
ligand-dependent nuclear receptor binding
protein domain specific binding
estrogen receptor binding
nuclear hormone receptor binding
histone deacetylase binding
peroxisome proliferator activated receptor binding
transcription regulatory region DNA binding
retinoid X receptor binding
thyroid hormone receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-hjwgp:80/10.239.166.163:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0