Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rilpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021430.2 | NP_067405.1 | 4 | 1007 | ||
GenBank mRNA | AB041584.1 | - | 4 | 637 | ||
AK205976.1 | - | 1 | 30 | |||
BC029965.1 | - | 4 | 685 | |||
BC051945.1 | - | 4 | 1007 | 5716540 | ||
BC051946.1 | - | 4 | 937 | 6403995 | ||
BC058384.1 | - | 4 | 900 | 5718399 |