Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006335.2 | NP_006326.1 | 3 | 192 | ||
GenBank mRNA | AF106622.1 | - | 3 | 163 | ||
AK315602.1 | - | 3 | 180 | |||
AU123184.1 | - | 3 | 192 | |||
BC000294.2 | - | 3 | 162 | 2819455 | ||
BC004439.1 | - | 3 | 162 | 2819455 | ||
BC007106.1 | - | 3 | 173 | 4284092 | ||
BC009784.1 | - | 3 | 184 | 3683125 | ||
BC013823.1 | - | 3 | 151 | |||
BC015098.1 | - | 3 | 190 | 3686369 | ||
BC016817.1 | - | 1 | 47 | |||
BC020833.1 | - | 3 | 170 | 4773149 | ||
X97544.1 | - | 3 | 169 |