Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aplf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170489.1 | NP_001163960.1 | 2, 3 | 374 | ||
NM_024251.4 | NP_077213.2 | 2, 3 | 244 | |||
GenBank mRNA | AK008513.1 | - | 2, 3 | 375 | ||
AK034191.1 | - | 2, 3 | 224 | |||
AK078805.1 | - | 2, 3 | 336 | |||
AK192630.1 | - | 2, 3 | 81 | |||
AK214829.1 | - | 1, 2 | 76 | |||
AK217392.1 | - | 2, 3 | 81 | |||
AK219060.1 | - | 2, 3 | 81 |