Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rprd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291134.1 | NP_001278063.1 | 5 | 940 | ||
NM_001291135.1 | NP_001278064.1 | 5 | 940 | |||
NM_001291136.1 | NP_001278065.1 | 4 | 807 | |||
NM_027434.3 | NP_081710.1 | 5 | 937 | |||
GenBank mRNA | AK011979.1 | - | 5 | 938 | ||
AK045404.1 | - | 4 | 613 | |||
AK139995.1 | - | 639 | ||||
AK147718.1 | - | 5 | 910 | |||
AK147930.1 | - | 5 | 899 | |||
AK152125.1 | - | 5 | 738 | |||
AK152220.1 | - | 5 | 738 | |||
BC028819.1 | - | 5 | 722 | 5100766 | ||
BC042447.1 | - | 5 | 649 | 5357316 | ||
BC054350.1 | - | 5 | 640 | 3488763 | ||
DQ372939.1 | - | 5 | 941 |