Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6ap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027439.4 | NP_081715.1 | 9 | 1668 | ||
GenBank mRNA | AK017482.1 | - | 9 | 1600 | ||
AK029405.1 | - | 9 | 1572 | |||
AK076495.1 | - | 9 | 1613 | |||
AK168139.1 | - | 9 | 1625 | |||
BC014706.1 | - | 9 | 1539 | 4457079 |