Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem230 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141971.1 | NP_001135443.1 | 4 | 1136 | ||
NM_027478.3 | NP_081754.2 | 4 | 1196 | |||
GenBank mRNA | AK017729.1 | - | 4 | 1190 | ||
AK170062.1 | - | 4 | 1169 | |||
BC080755.1 | - | 3 | 2560 | |||
BY388388.1 | - | 1 | 17 |