Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Anapc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291348.1 | NP_001278277.1 | 6 | 664 | ||
NM_001291349.1 | NP_001278278.1 | 5 | 614 | |||
NM_001291352.1 | NP_001278281.1 | 6 | 657 | |||
NM_001291353.1 | NP_001278282.1 | 6 | 657 | |||
NM_027532.4 | NP_081808.2 | 6 | 699 | |||
GenBank mRNA | AK014309.1 | - | 6 | 616 | ||
AK135201.1 | - | 6 | 615 | |||
AK139635.1 | - | 6 | 656 | |||
BC006736.1 | - | 6 | 657 | 3964006 | ||
BC080730.1 | - | 5 | 758 | 6507829 | ||
DV659149.1 | - | 6 | 669 |