Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afmid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027827.3 | NP_082103.1 | 11 | 1398 | ||
GenBank mRNA | AF399717.1 | - | 11 | 1393 | ||
AY099479.1 | - | 11 | 1392 | |||
BC043309.1 | - | 11 | 1364 | |||
BC066780.1 | - | 11 | 1377 | 30301832 |