Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Telo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163661.1 | NP_001157133.1 | 16 | 2519 | ||
NM_027880.2 | NP_082156.2 | 16 | 2430 | |||
GenBank mRNA | AK004582.1 | - | 16 | 2300 | ||
AK028658.1 | - | 16 | 2284 | |||
AK045321.1 | - | 9 | 1966 | |||
AK050773.1 | - | 16 | 2249 | |||
AK129189.1 | - | 15 | 2592 | |||
AK143827.1 | - | 16 | 2520 | |||
AK146963.1 | - | 16 | 2306 | |||
AK154727.1 | - | 16 | 2332 | |||
BC011077.1 | - | 16 | 2280 | 3984911 | ||
BC080713.1 | - | 15 | 2539 |