Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028048.2 | NP_082324.1 | 3 | 1001 | ||
GenBank mRNA | AK007453.1 | - | 3 | 1003 | ||
AK035739.1 | - | 972 | ||||
AK157873.1 | - | 2 | 1389 | |||
AK171131.1 | - | 3 | 1008 | |||
BC004569.1 | - | 3 | 441 | |||
BC019996.1 | - | 3 | 943 | 4207271 |