Gene Symbol: Tmem68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028097.3 | NP_082373.1 | 4 | 631 | ||
GenBank mRNA | AK008493.1 | - | 4 | 665 | ||
AK048210.1 | - | 4 | 631 | |||
AK084881.1 | - | 4 | 665 | |||
AK088600.1 | - | 4 | 674 | |||
AK147765.1 | - | 4 | 664 | |||
AK162563.1 | - | 4 | 664 | |||
AK165916.1 | - | 4 | 693 | |||
AK166237.1 | - | 4 | 717 | |||
BC016240.1 | - | 4 | 649 | 4506688 |