Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc71l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162903.1 | NP_001156375.1 | 1 | 1366 | ||
GenBank mRNA | AK008367.1 | - | 1 | 510 | ||
AK171109.1 | - | 1 | 1016 | |||
BC028641.1 | - | 2 | 659 | |||
BC037374.1 | - | 1 | 226 | |||
BC049684.1 | - | 1 | 406 | |||
BC061478.1 | - | 1 | 737 |