Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028243.3 | NP_082519.1 | 4 | 535 | ||
GenBank mRNA | AK011112.1 | - | 4 | 517 | ||
AK051677.1 | - | 4 | 492 | |||
AK054462.1 | - | 4 | 473 | |||
AK166282.1 | - | 4 | 492 | |||
BC026424.1 | - | 4 | 331 | |||
BC055022.1 | - | 4 | 468 | 4017462 | ||
BC089575.1 | - | 4 | 490 |