Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011832.1 | - | 2 | 119 | ||
AK030398.1 | - | 2 | 318 | |||
AK047429.1 | - | 1 | 101 | |||
AK134257.1 | - | 2 | 118 | |||
AK135723.1 | - | 1 | 102 | |||
AK162903.1 | - | 1 | 541 | |||
AY092766.1 | - | 1 | 210 | |||
BC023762.1 | - | 2 | 388 | |||
BC057991.1 | - | 2 | 159 | 5323559 | ||
BE994245.1 | - | 1 | 260 | |||
EF410128.1 | - | 343 |