Gene Symbol: Ppp2r2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028392.3 | NP_082668.1 | 1, 2 | 318 | ||
GenBank mRNA | AF536771.1 | - | 1, 2 | 87 | ||
AK013600.1 | - | 1, 2 | 320 | |||
AK018119.1 | - | 1, 2 | 275 | |||
AK039592.1 | - | 1, 2 | 161 | |||
AK079059.1 | - | 768 | ||||
AK148956.1 | - | 1, 2 | 289 | |||
BC026686.1 | - | 1, 2 | 256 | 5065516 | ||
BC066007.1 | - | 1, 2 | 558 | |||
BC085147.1 | - | 1, 2 | 470 | |||
BC088979.1 | - | 1, 2 | 253 | 5690600 |