Gene Symbol: Ppp2r2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028392.3 | NP_082668.1 | 5 | 832 | ||
GenBank mRNA | AF512670.1 | - | 5 | 538 | ||
AF536771.1 | - | 5 | 601 | |||
AK013600.1 | - | 5 | 834 | |||
AK018119.1 | - | 5 | 789 | |||
AK039592.1 | - | 5 | 675 | |||
AK079059.1 | - | 1282 | ||||
AK148956.1 | - | 5 | 803 | |||
BC066007.1 | - | 5 | 1072 | |||
BC085147.1 | - | 5 | 985 | |||
BC088979.1 | - | 5 | 767 | 5690600 |