Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028554.3 | NP_082830.2 | 3 | 491 | ||
GenBank mRNA | AK006935.1 | - | 3 | 492 |