Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028724.4 | NP_083000.5 | 5, 6 | 609 | ||
GenBank mRNA | AK150680.1 | - | 3, 4 | 417 | ||
AK151332.1 | - | 2, 3 | 373 | |||
AK152893.1 | - | 3, 4 | 417 | |||
BC040390.1 | - | 5, 6 | 603 | 3156682 | ||
CN461821.1 | - | 4, 5 | 593 |