Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028724.4 | NP_083000.5 | 13 | 4111 | ||
GenBank mRNA | AK014548.1 | - | 5 | 2582 | ||
AK040763.1 | - | 5 | 2583 | |||
AK150680.1 | - | 11 | 3918 | |||
AK151332.1 | - | 11 | 3875 | |||
AK152025.1 | - | 6 | 2556 | |||
AK152893.1 | - | 11 | 3919 | |||
AK154421.1 | - | 5 | 2528 | |||
AK163047.1 | - | 5 | 3096 | |||
AK171064.1 | - | 5 | 2496 | |||
BC005529.1 | - | 5 | 2543 | 3495681 | ||
BC040390.1 | - | 13 | 4103 | 3156682 |