Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164362.1 | NP_001157834.1 | 4 | 889 | ||
NM_028293.1 | NP_082569.1 | 3 | 513 | |||
NM_028760.2 | NP_083036.2 | 4 | 865 | |||
GenBank mRNA | AK004655.1 | - | 4 | 891 | ||
AK012311.1 | - | 4 | 662 | |||
AK028216.1 | - | 3 | 499 | |||
AK088548.1 | - | 4 | 572 | |||
BC026966.1 | - | 1 | 221 | |||
BC031396.1 | - | 4 | 818 | 4950530 |