Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Armc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028768.3 | NP_083044.2 | 19 | 2026 | ||
GenBank mRNA | AK004793.2 | - | 19 | 2027 | ||
BC030311.1 | - | 1 | 178 |