Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sash3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028773.3 | NP_083049.1 | 8 | 2528 | ||
GenBank mRNA | AK004734.2 | - | 8 | 2529 | ||
AK156202.1 | - | 8 | 2593 | |||
AK171017.1 | - | 8 | 2526 | |||
BC028773.1 | - | 5 | 2090 | |||
BC064734.1 | - | 8 | 2513 |