Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec14l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166506.1 | NP_001159978.1 | 13, 14 | 1818 | ||
NM_001166507.1 | NP_001159979.1 | 13, 14 | 1818 | |||
NM_028777.3 | NP_083053.2 | 13, 14 | 1818 | |||
GenBank mRNA | AK004818.1 | - | 13, 14 | 1743 | ||
AK172947.1 | - | 10, 11 | 1353 | |||
AK195036.1 | - | 1, 2 | 128 | |||
AK200242.1 | - | 1, 2 | 128 | |||
BC005766.1 | - | 13, 14 | 1776 | 2649054 | ||
BC086620.1 | - | 13, 14 | 1751 |