Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc3l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289487.1 | NP_001276416.1 | 5 | 1550 | ||
NM_001289488.1 | NP_001276417.1 | 5 | 1301 | |||
NM_001289489.1 | NP_001276418.1 | 4 | 1196 | |||
NM_028807.4 | NP_083083.3 | 4 | 1445 | |||
GenBank mRNA | AK004674.1 | - | 5 | 1302 | ||
AK005443.1 | - | 3 | 1192 | |||
AK154875.1 | - | 5 | 1964 | |||
BC042799.1 | - | 1 | 185 | |||
BC075664.1 | - | 4 | 1286 | 30604610 |