Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Snx16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127191.2 | NP_001120663.1 | 4 | 804 | ||
NM_029068.4 | NP_083344.3 | 4 | 804 | |||
GenBank mRNA | AK015872.1 | - | 4 | 765 | ||
AK032055.1 | - | 4 | 761 | |||
AK166422.1 | - | 4 | 778 | |||
BC019424.1 | - | 3 | 569 | 5052234 | ||
BC070431.1 | - | 747 |